4-Chloro-3-iodotoluene ≥97%

Supplier: AOBChem USA

Warning

23959-25G 23959-5G
76928-252EA 453.01 USD
76928-252 76928-254
4-Chloro-3-iodotoluene ≥97%
4-Chloro-3-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₆ClI
Storage Temperature: Ambient
MDL Number: MFCD09753727
CAS Number: 2401-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR