4-Fluoro-3-phenylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

23929-1G 23929-25G 23929-500MG 23929-5G
76928-236EA 201.58 USD
76928-236 76928-238 76928-240 76928-242
4-Fluoro-3-phenylbenzaldehyde ≥97%
4-Fluoro-3-phenylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₉FO
MW: 200.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09042262
CAS Number: 926221-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR