4-Bromo-N-phenylbenzamide ≥97%

Supplier: AOBChem USA

Warning

23885-25G 23885-5G
76928-224EA 439.22 USD
76928-224 76928-226
4-Bromo-N-phenylbenzamide ≥97%
4-Bromo-N-phenylbenzamide

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrNO
MW: 276.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00624477
CAS Number: 6846-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR