4-Formyl-2,6-dimethylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

23852-1G 23852-25G 23852-500MG 23852-5G
76928-212EA 147.47 USD
76928-212 76928-214 76928-216 76928-218
4-Formyl-2,6-dimethylphenylboronic acid pinacol ester ≥97%
4-Formyl-2,6-dimethylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₃
MW: 260.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30344662
CAS Number: 1951423-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR