5-Chloro-2-fluoro-3-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

23838-1G 23838-25G 23838-500MG 23838-5G
76928-196EA 225.98 USD
76928-196 76928-198 76928-200 76928-202
5-Chloro-2-fluoro-3-(trifluoromethyl)phenylboronic acid ≥97%
5-Chloro-2-fluoro-3-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₄BClF₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD28806015
CAS Number: 2121513-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR