4'-Chloro-[1,1'-biphenyl]-2-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

23817-1G 23817-25G 23817-500MG 23817-5G
76928-184EA 228.1 USD
76928-184 76928-186 76928-188 76928-190
4'-Chloro-[1,1'-biphenyl]-2-carboxaldehyde ≥97%
4'-Chloro-[1,1'-biphenyl]-2-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₉ClO
Storage Temperature: Ambient
MDL Number: MFCD03424629
CAS Number: 153850-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR