4-Bromo-2-fluoro-5-methylphenol ≥97%

Supplier: AOBChem USA

Warning

23663-25G 23663-5G
76928-148EA 595.17 USD
76928-148 76928-150
4-Bromo-2-fluoro-5-methylphenol ≥97%
4-Bromo-2-fluoro-5-methylphenol
Formula: C₇H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD09261252
CAS Number: 550400-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR