4-Fluoro-3-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

23511-25G 23511-5G
76928-090EA 488.02 USD
76928-090 76928-092
4-Fluoro-3-hydroxybenzaldehyde ≥97%
4-Fluoro-3-hydroxybenzaldehyde
Formula: C₇H₅FO₂
Storage Temperature: Ambient
MDL Number: MFCD11110190
CAS Number: 103438-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR