4-Iodo-2-(morpholino)pyridine ≥97%

Supplier: AOBChem USA

Warning

23506-1G 23506-25G 23506-500MG 23506-5G
76928-076EA 173.99 USD
76928-076 76928-078 76928-080 76928-082
4-Iodo-2-(morpholino)pyridine ≥97%
4-Iodo-2-(morpholino)pyridine

Precaución: for Research Use Only

Formula: C₉H₁₁IN₂O
MW: 290.101 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22056770
CAS Number: 1370025-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR