4-Fluoro-3-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Fluoro-3-methoxyphenyl)methanol

23423-25G 23423-5G
76928-038EA 254.62 USD
76928-038 76928-040
4-Fluoro-3-methoxybenzyl alcohol ≥97%
4-Fluoro-3-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉FO₂
MW: 156.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272133
CAS Number: 128495-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR