4-Iodo-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

23365-25G 23365-5G
76928-028EA 339.49 USD
76928-028 76928-030
4-Iodo-2-methoxypyridine ≥97%
4-Iodo-2-methoxypyridine
Formula: C₆H₆INO
Storage Temperature: Ambient
MDL Number: MFCD08277272
CAS Number: 98197-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR