5-Chloro-2,3-dimethoxypyridine ≥97% research grade

Supplier: AOBChem USA

Warning

23333-1G 23333-25G 23333-5G
76928-018EA 62.6 USD
76928-018 76928-020 76928-022
5-Chloro-2,3-dimethoxypyridine ≥97% research grade
5-Chloro-2,3-dimethoxypyridine
Formula: C₇H₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD09972187
CAS Number: 284040-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR