4-Bromo-2-iodo-6-methylaniline ≥97%

Supplier: AOBChem USA

Warning

23303-1G 23303-25G 23303-500MG 23303-5G
76928-004EA 203.7 USD
76928-004 76928-006 76928-008 76928-010
4-Bromo-2-iodo-6-methylaniline ≥97%
4-Bromo-2-iodo-6-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrIN
Storage Temperature: Ambient
MDL Number: MFCD16659506
CAS Number: 922170-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR