4-Bromo-6-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

23279-1G 23279-25G 23279-500MG 23279-5G
76927-976EA 159.14 USD
76927-976 76927-978 76927-980 76927-982
4-Bromo-6-methyl-1H-indazole ≥97%
4-Bromo-6-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.059 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07781508
CAS Number: 885521-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR