1-(4-Bromo-2-fluorobenzyl)piperidine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-fluorophenyl-1-(piperidinomethyl)benzen

23221-1G 23221-25G 23221-5G
76927-954EA 64.72 USD
76927-954 76927-956 76927-958
1-(4-Bromo-2-fluorobenzyl)piperidine ≥97%
1-(4-Bromo-2-fluorobenzyl)piperidine

Precaución: for Research Use Only

Formula: C₁₂H₁₅BrFN
MW: 272.157 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14645209
CAS Number: 1200131-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR