4-Iodo-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

23191-1G 23191-25G 23191-500MG 23191-5G
76927-942EA 120.95 USD
76927-942 76927-944 76927-946 76927-948
4-Iodo-1H-indazole ≥97%
4-Iodo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅IN₂
Storage Temperature: Refrigerator
MDL Number: MFCD07781607
CAS Number: 885522-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR