4-Ethoxy-2,6-difluorophenol ≥97%

Supplier: AOBChem USA

Warning

22863-1G 22863-25G 22863-5G
76927-910EA 76.39 USD
76927-910 76927-912 76927-914
4-Ethoxy-2,6-difluorophenol ≥97%
4-Ethoxy-2,6-difluorophenol

Precaución: for Research Use Only

Formula: C₈H₈F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09258699
CAS Number: 1017779-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR