5-Bromo-2,3-difluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

22743-1G 22743-25G 22743-500MG 22743-5G
76927-824EA 287.51 USD
76927-824 76927-826 76927-828 76927-830
5-Bromo-2,3-difluorobenzyl alcohol ≥97%
5-Bromo-2,3-difluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅BrF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD04972473
CAS Number: 887585-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR