5-Bromo-N,N-dimethylpyridin-3-amine ≥97%

Supplier: AOBChem USA

Warning

22736-1G 22736-25G 22736-5G
76927-814EA 64.72 USD
76927-814 76927-816 76927-818
5-Bromo-N,N-dimethylpyridin-3-amine ≥97%
5-Bromo-N,N-dimethylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD12828170
CAS Number: 342602-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR