5-Chloro-2-iodo-1-methyl-3-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

22697-1G 22697-25G 22697-5G
76927-800EA 93.36 USD
76927-800 76927-802 76927-804
5-Chloro-2-iodo-1-methyl-3-nitrobenzene ≥97%
5-Chloro-2-iodo-1-methyl-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅ClINO₂
MW: 297.478 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12024407
CAS Number: 1150617-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR