5-Bromo-4-chloro-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

22530-25G 22530-5G
76927-754EA 268.41 USD
76927-754 76927-756
5-Bromo-4-chloro-1H-indazole ≥97%
5-Bromo-4-chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.4771 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11007933
CAS Number: 1082041-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR