5-Bromo-2-butoxy-1,3-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

22476-1G 22476-25G 22476-5G
76927-714EA 64.72 USD
76927-714 76927-716 76927-718
5-Bromo-2-butoxy-1,3-difluorobenzene ≥97%
5-Bromo-2-butoxy-1,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD18917756
CAS Number: 1309932-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR