5-Bromo-1,3-difluoro-2-isopropoxybenzene ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
Synonyms:
5-Bromo-1,3-difluoro-2-(propan-2-yloxy)benzene
22472-1G
22472-25G
22472-5G
76927-702EA
62.6
USD
76927-702
76927-704
76927-706
5-Bromo-1,3-difluoro-2-isopropoxybenzene ≥97% research grade
5-Bromo-1,3-difluoro-2-isopropoxybenzene
Formula:
C₉H₉BrF₂O MW: 251.06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18917884 CAS Number: 1309933-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...