4-Hydroxy-3-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

22373-25G 22373-5G
76927-678EA 477.41 USD
76927-678 76927-680
4-Hydroxy-3-methoxyphenylboronic acid pinacol ester ≥97%
4-Hydroxy-3-methoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₉BO₄
MW: 250.099 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093723
CAS Number: 269410-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR