4-Cyclopropylbenzonitrile ≥97%

Supplier: AOBChem USA

Warning

22306-1G 22306-25G 22306-500MG 22306-5G
76927-664EA 272.66 USD
76927-664 76927-666 76927-668 76927-670
4-Cyclopropylbenzonitrile ≥97%
4-Cyclopropylbenzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₉N
Storage Temperature: Refrigerator
MDL Number: MFCD06802400
CAS Number: 1126-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR