4-Bromo-3-(trifluoromethyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(4-Bromo-3-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

22259-1G 22259-25G 22259-500MG 22259-5G
76927-638EA 155.96 USD
76927-638 76927-640 76927-642 76927-644
4-Bromo-3-(trifluoromethyl)phenylboronic acid pinacol ester ≥97%
4-Bromo-3-(trifluoromethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BBrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD18383430
CAS Number: 1256944-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR