4-Methoxy-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

22094-1G 22094-25G 22094-5G
76927-606EA 94.43 USD
76927-606 76927-608 76927-610
4-Methoxy-1H-indazole ≥97%
4-Methoxy-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD09261131
CAS Number: 351210-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR