Methyl 5-bromo-6-methylpicolinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-6-methylpyridine-2-carboxylic acid methyl ester

21987-1G 21987-25G 21987-500MG 21987-5G
76927-552EA 127.31 USD
76927-552 76927-554 76927-556 76927-558
Methyl 5-bromo-6-methylpicolinate ≥97%
Methyl 5-bromo-6-methylpicolinate
Formula: C₈H₈BrNO₂
MW: 230.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09753045
CAS Number: 1215860-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR