4-Bromo-4'-isopropyloxybiphenyl ≥97%

Supplier: AOBChem USA

Warning

21962-25G 21962-5G
76927-544EA 240.83 USD
76927-544 76927-546
4-Bromo-4'-isopropyloxybiphenyl ≥97%
4-Bromo-4'-isopropyloxybiphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrO
Storage Temperature: Ambient
MDL Number: MFCD19105396
CAS Number: 153486-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR