4-Chloro-3-(trifluoromethoxy)bromobenzene ≥97%

Supplier: AOBChem USA

Warning

21883-25G 21883-5G
76927-534EA 300.24 USD
76927-534 76927-536
4-Chloro-3-(trifluoromethoxy)bromobenzene ≥97%
4-Chloro-3-(trifluoromethoxy)bromobenzene
Formula: C₇H₃BrClF₃O
Storage Temperature: Ambient
MDL Number: MFCD09878140
CAS Number: 406232-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR