4-Carbamoyl-2-methoxyphenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
21869-1G
21869-25G
21869-500MG
21869-5G
76927-522EA
195.21
USD
76927-522
76927-524
76927-526
76927-528
4-Carbamoyl-2-methoxyphenylboronic acid ≥97%
4-Carbamoyl-2-methoxyphenylboronic acid
Precaución: for Research Use Only
Formula:
C₈H₁₀BNO₄ MW: 194.98 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19105392 CAS Number: 1451392-06-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...