5-Bromo-2-fluoro-3-trifluoromethylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

21617-25G 21617-5G
76927-476EA 381.93 USD
76927-476 76927-478
5-Bromo-2-fluoro-3-trifluoromethylphenylboronic acid ≥97%
5-Bromo-2-fluoro-3-trifluoromethylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₄BBrF₄O₂
MW: 286.814 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18311845
CAS Number: 2096341-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR