5-Chloro-2-fluoro-3-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

21407-1G 21407-25G 21407-500MG 21407-5G
76927-434EA 155.96 USD
76927-434 76927-436 76927-438 76927-440
5-Chloro-2-fluoro-3-methoxypyridine ≥95%
5-Chloro-2-fluoro-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅ClFNO
MW: 161.561 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16610383
CAS Number: 1227580-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR