4-Methyl-1H-indazole-5-boronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (4-Methyl-1H-indazol-5-yl)boronic acid

21383-1G 21383-25G 21383-5G
76927-420EA 93.36 USD
76927-420 76927-422 76927-424
4-Methyl-1H-indazole-5-boronic acid ≥95%
4-Methyl-1H-indazole-5-boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₂
MW: 175.9800 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12028644
CAS Number: 1245816-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR