5-Chloro-3-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Chloropyridine-3-boronic acid Pinacol ester, 2-(5-Chloro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

21167-25G 21167-5G
76927-384EA 421.18 USD
76927-384 76927-386
5-Chloro-3-pyridineboronic acid pinacol ester ≥97%
5-Chloro-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₅BClNO₂
Storage Temperature: Ambient
MDL Number: MFCD06798284
CAS Number: 865186-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR