4-Bromo-3-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Bromo-3-methylphenyl)methanol

21161-25G 21161-5G
76927-372EA 488.02 USD
76927-372 76927-374
4-Bromo-3-methylbenzyl alcohol ≥97%
4-Bromo-3-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD09264005
CAS Number: 149104-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR