4-Chloro-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

21159-25G 21159-5G
76927-364EA 183.54 USD
76927-364 76927-366
4-Chloro-2-methylbenzaldehyde ≥97%
4-Chloro-2-methylbenzaldehyde
Formula: C₈H₇ClO
Storage Temperature: Ambient
MDL Number: MFCD02261735
CAS Number: 40137-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR