5-Bromobenzo[d][1,3]dioxole-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-1,3-benzodioxole-4-carboxaldehyde

21068-1G 21068-25G 21068-5G
76927-334EA 62.6 USD
76927-334 76927-336 76927-338
5-Bromobenzo[d][1,3]dioxole-4-carbaldehyde ≥97%
5-Bromobenzo[d][1,3]dioxole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅BrO₃
MW: 229.027 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02069012
CAS Number: 72744-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR