4-Bromo-2-fluoro-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

21027-1G 21027-25G 21027-5G
76927-308EA 112.46 USD
76927-308 76927-310 76927-312
4-Bromo-2-fluoro-5-methylbenzaldehyde ≥97%
4-Bromo-2-fluoro-5-methylbenzaldehyde
Formula: C₈H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD07782048
CAS Number: 916792-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR