5-Bromo-2-fluoro-3-methylbenzonitrile ≥97%

Supplier: AOBChem USA

Warning

21014-1G 21014-25G 21014-500MG 21014-5G
76927-298EA 187.78 USD
76927-298 76927-300 76927-302 76927-304
5-Bromo-2-fluoro-3-methylbenzonitrile ≥97%
5-Bromo-2-fluoro-3-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFN
Storage Temperature: Ambient
MDL Number: MFCD25955533
CAS Number: 1110502-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR