4-Bromo-2-fluoro-5-methylpyridine ≥95%

Supplier: AOBChem USA

Warning

20834-1G 20834-25G 20834-500MG 20834-5G
76927-250EA 138.98 USD
76927-250 76927-252 76927-254 76927-256
4-Bromo-2-fluoro-5-methylpyridine ≥95%
4-Bromo-2-fluoro-5-methylpyridine
Formula: C₆H₅BrFN
Storage Temperature: Refrigerator
MDL Number: MFCD16609965
CAS Number: 1227577-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR