5-Chloro-2-fluoro-4-pyridinecarboxylic acid ≥97%

Supplier: AOBChem USA

Warning

20714-1G 20714-25G 20714-500MG 20714-5G
76927-220EA 167.63 USD
76927-220 76927-222 76927-224 76927-226
5-Chloro-2-fluoro-4-pyridinecarboxylic acid ≥97%
5-Chloro-2-fluoro-4-pyridinecarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₂
Storage Temperature: Refrigerator
CAS Number: 1211532-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR