4-Fluoro-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoroindazole

20630-25G 20630-5G
76927-202EA 198.39 USD
76927-202 76927-204
4-Fluoro-1H-indazole ≥97%
4-Fluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅FN₂
MW: 136.1264 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07371561
CAS Number: 341-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR