5-(Cyclopropylcarbamoyl)-2-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

20595-1G 20595-25G 20595-500MG 20595-5G
76927-170EA 177.18 USD
76927-170 76927-172 76927-174 76927-176
5-(Cyclopropylcarbamoyl)-2-fluorophenylboronic acid ≥97%
5-(Cyclopropylcarbamoyl)-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BFNO₃
MW: 223.009 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11855834
CAS Number: 874289-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR