5-(Diethylcarbamoyl)-2-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (5-(Diethylcarbamoyl)-2-fluorophenyl)boronic acid

20584-1G 20584-25G 20584-500MG 20584-5G
76927-154EA 132.62 USD
76927-154 76927-156 76927-158 76927-160
5-(Diethylcarbamoyl)-2-fluorophenylboronic acid ≥97%
5-(Diethylcarbamoyl)-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BFNO₃
MW: 239.0500 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08436045
CAS Number: 874289-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR