5-(Dimethylcarbamoyl)-2-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-(Dimethylcarbamoyl)-2-fluorobenzeneboronic acid, (5-(Dimethylcarbamoyl)-2-fluorophenyl)boronic acid, 5-Dimethylcarbamoyl-2-fluorobenzeneboronic acid

20580-1G 20580-25G 20580-5G
76927-140EA 79.57 USD
76927-140 76927-142 76927-144
5-(Dimethylcarbamoyl)-2-fluorophenylboronic acid ≥97%
5-(Dimethylcarbamoyl)-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BFNO₃
MW: 211.00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08235102
CAS Number: 874289-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR