4-Bromo-2-fluoro-5-methoxypyridine ≥97%

Supplier: AOBChem USA

20460-1G 20460-25G 20460-500MG 20460-5G
76927-104EA 289.63 USD
76927-104 76927-106 76927-108 76927-110
4-Bromo-2-fluoro-5-methoxypyridine ≥97%
4-Bromo-2-fluoro-5-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFNO
MDL Number: MFCD16609958
CAS Number: 1227564-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR