4-Fluoro-3-isopropoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoro-3-(1-methylethoxy)-benzaldehyde

20444-1G 20444-25G 20444-5G
76927-090EA 117.76 USD
76927-090 76927-092 76927-094
4-Fluoro-3-isopropoxybenzaldehyde ≥95%
4-Fluoro-3-isopropoxybenzaldehyde
Formula: C₁₀H₁₁FO₂
MW: 182.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18447608
CAS Number: 1236365-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR