4-Fluoro-3-isopropoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

20324-1G 20324-25G 20324-5G
76927-072EA 102.91 USD
76927-072 76927-074 76927-076
4-Fluoro-3-isopropoxyphenylboronic acid ≥97%
4-Fluoro-3-isopropoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BFO₃
Storage Temperature: Ambient
MDL Number: MFCD18447603
CAS Number: 1451390-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR