4-Bromo-3,5-dichloroaniline ≥97%

Supplier: AOBChem USA

Warning

20212-25G 20212-5G
76927-056EA 381.93 USD
76927-056 76927-058
4-Bromo-3,5-dichloroaniline ≥97%
4-Bromo-3,5-dichloroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrCl₂N
MW: 240.9100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878195
CAS Number: 1940-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR